This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AZIN1 SIRNA Assays ›
Gene Symbol:
Azin1
Gene Name:
antizyme inhibitor 1
Gene Aliases:
Oazi, Oazin
Chromosome Location:
Chr.7: 77345671 - 77372393 on Build Rnor_6.0
UniGene ID:
Rn.6290
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 52092

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Azin1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301702.1 NP_001288631.1 5, 6 1082
NM_022585.2 NP_072107.2 6, 7 1240
XM_006241568.2 XP_006241630.1 6, 7 1213
XM_006241569.2 XP_006241631.1 5, 6 1077
XM_008765457.1 XP_008763679.1 5, 6 1077
GenBank mRNA BC061550.1 - 3, 4 478 6918922
D50734.1 - 5, 6 875
D89983.1 - 6, 7 1188

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_148174, NM_015878, BT006858, BC042404, BC013420, and others.
This siRNA also targets the following transcripts in Mouse: NM_001102458, NM_018745, AK088671, BC043722, AK049680, and others.

Gene Ontology Categories:

Function(s) Process(es)

polyamine biosynthetic process
negative regulation of protein catabolic process
positive regulation of catalytic activity
negative regulation of catalytic activity
positive regulation of polyamine transmembrane transport
catalytic activity
ornithine decarboxylase inhibitor activity
ornithine decarboxylase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm