This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AZIN1 SIRNA Assays ›
Gene Symbol:
Azin1
Gene Name:
antizyme inhibitor 1
Gene Aliases:
Oazi, Oazin
Chromosome Location:
Chr.7: 77345671 - 77372393 on Build Rnor_6.0
UniGene ID:
Rn.6290
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 51997

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Azin1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301702.1 NP_001288631.1 5 1063
NM_022585.2 NP_072107.2 6 1221
XM_006241568.2 XP_006241630.1 6 1194
XM_006241569.2 XP_006241631.1 5 1058
XM_008765457.1 XP_008763679.1 5 1058
GenBank mRNA BC061550.1 - 3 459 6918922
D50734.1 - 5 856
D89983.1 - 6 1169

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

polyamine biosynthetic process
negative regulation of protein catabolic process
positive regulation of catalytic activity
negative regulation of catalytic activity
positive regulation of polyamine transmembrane transport
catalytic activity
ornithine decarboxylase inhibitor activity
ornithine decarboxylase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0