This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARF1 SIRNA Assays ›
Gene Symbol:
Arf1
Gene Name:
ADP-ribosylation factor 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 45562700 - 45579192 on Build Rnor_6.0
UniGene ID:
Rn.93735
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 51967

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022518.3 NP_071963.1 2 167
XM_006246525.2 XP_006246587.1 2 214
XM_008767870.1 XP_008766092.1 3 259
GenBank mRNA BC061552.1 - 2 183 6889332
L12380.1 - 2 167

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007476, NM_001130408, BC031986, AK168686, D87898, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of receptor internalization
cellular copper ion homeostasis
actin filament organization
small GTPase mediated signal transduction
protein transport
vesicle-mediated transport
regulation of Arp2/3 complex-mediated actin nucleation
very-low-density lipoprotein particle assembly
positive regulation of catalytic activity
positive regulation of endocytosis
positive regulation of calcium ion-dependent exocytosis
positive regulation of protein secretion
Golgi to transport vesicle transport
long term synaptic depression
positive regulation of dendritic spine development
synaptic vesicle budding
dendritic spine organization
lysosomal membrane organization
positive regulation of sodium ion transmembrane transport
positive regulation of late endosome to lysosome transport
positive regulation of ER to Golgi vesicle-mediated transport
regulation of phospholipid metabolic process
magnesium ion binding
protein binding
GTP binding
GDP binding
protein homodimerization activity
poly(A) RNA binding
phospholipase D activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0