This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HNF4A SIRNA Assays ›
Gene Symbol:
Hnf4a
Gene Name:
hepatocyte nuclear factor 4, alpha
Gene Aliases:
Hnf4, Hnf4alpha
Chromosome Location:
Chr.3: 159902441 - 159965003 on Build Rnor_6.0
UniGene ID:
Rn.12238
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 51661

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hnf4a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270931.1 NP_001257860.1 3 454
NM_001270933.1 NP_001257862.1 3 454
NM_022180.2 NP_071516.2 2 414
XM_006235515.2 XP_006235577.1 2 409
XM_006235516.2 XP_006235578.1 2 409
GenBank mRNA CK483260.1 - 2 243
D10554.1 - 2 290
EF193390.1 - 2 235
EF193392.1 - 2 216
FJ608816.1 - 3 454
FJ608817.1 - 3 454
FJ608818.1 - 3 454
FM042560.1 - 2 419
X57133.1 - 2 305

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008261, FJ608819, AK143948, BC039220, FJ608820, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
ornithine metabolic process
lipid metabolic process
acyl-CoA metabolic process
xenobiotic metabolic process
establishment of tissue polarity
sex differentiation
blood coagulation
negative regulation of cell proliferation
response to glucose
regulation of gastrulation
regulation of lipid metabolic process
signal transduction involved in regulation of gene expression
cell differentiation
negative regulation of cell growth
intracellular receptor signaling pathway
regulation of microvillus assembly
negative regulation of protein import into nucleus, translocation
response to drug
negative regulation of tyrosine phosphorylation of Stat5 protein
glucose homeostasis
steroid hormone mediated signaling pathway
negative regulation of sequence-specific DNA binding transcription factor activity
cell-cell junction organization
positive regulation of gluconeogenesis
positive regulation of fatty acid biosynthetic process
positive regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
regulation of insulin secretion
response to cAMP
lipid homeostasis
phospholipid homeostasis
SMAD protein signal transduction
triglyceride homeostasis
negative regulation of activation of JAK2 kinase activity
positive regulation of cholesterol homeostasis
fatty-acyl-CoA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
receptor binding
fatty acid binding
protein binding
transcription factor binding
zinc ion binding
palmitoyl-CoA hydrolase activity
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
repressing transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0