This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNV1 SIRNA Assays ›
Gene Symbol:
Kcnv1
Gene Name:
potassium channel, voltage-gated modifier subfamily V, member 1
Gene Aliases:
Kv8.1
Chromosome Location:
Chr.7: 84016864 - 84023205 on Build Rnor_6.0
UniGene ID:
Rn.42920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 51335

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnv1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021697.1 NP_067729.1 1, 2 488
GenBank mRNA FQ211837.1 - 499
X98564.1 - 1, 2 488

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_014379, AK313469, BC028739, AK297393, AF167082, and others.
This siRNA also targets the following transcripts in Mouse: NM_026200, AK139482, BC128478, AK012275, BC128477, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of ion transmembrane transport
protein homooligomerization
potassium ion transmembrane transport
delayed rectifier potassium channel activity
potassium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0