This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CNN3 SIRNA Assays ›
Gene Symbol:
Cnn3
Gene Name:
calponin 3, acidic
Gene Aliases:
-
Chromosome Location:
Chr.2: 225005069 - 225036179 on Build Rnor_6.0
UniGene ID:
Rn.57635
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 50935

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnn3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019359.1 NP_062232.1 2 329
GenBank mRNA BC062020.1 - 2 332 5623281
U06755.1 - 2 329

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

epithelial cell differentiation
actomyosin structure organization
negative regulation of ATPase activity
actin binding
calmodulin binding
microtubule binding
protein binding, bridging

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header