This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNN1 SIRNA Assays ›
Gene Symbol:
Kcnn1
Gene Name:
potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 1
Gene Aliases:
KCa2.1
Chromosome Location:
Chr.16: 20336784 - 20349168 on Build Rnor_6.0
UniGene ID:
Rn.44422
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 50901

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019313.1 NP_062186.1 3, 4 913
XM_006252933.2 XP_006252995.1 3, 4 946
GenBank mRNA AF000973.1 - 3, 4 913
U69885.1 - 3, 4 679

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

potassium ion transmembrane transport
calmodulin binding
small conductance calcium-activated potassium channel activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0