This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KXD1 SIRNA Assays ›
Gene Symbol:
Kxd1
Gene Name:
KxDL motif containing 1
Gene Aliases:
0610030B01Rik, 2810422J05Rik, C78305
Chromosome Location:
Chr.8: 70513396 - 70523180 on Build GRCm38
UniGene ID:
Mm.482129
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 508670

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kxd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029366.2 NP_083642.1 5 1059
GenBank mRNA AK013135.1 - 5 975
BC038552.1 - 4 952
BC048938.1 - 5 1058 6399490
BC053944.1 - 5 1058

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

vesicle-mediated transport
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0