This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 5830415F09RIK SIRNA Assays ›
Gene Symbol:
5830415F09Rik
Gene Name:
RIKEN cDNA 5830415F09 gene
Gene Aliases:
AV014846, Nap1
Chromosome Location:
Chr.4: 46376504 - 46393557 on Build GRCm38
UniGene ID:
Mm.45973
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 508455

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 5830415F09Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029086.2 NP_083362.1 5 1192
GenBank mRNA AK017931.1 - 5 1203
AK044073.1 - 5 1186
AK052584.1 - 5 1208
AK153576.1 - 5 1193
BC092543.1 - 5 1193 30315335

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
acyl-CoA hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0