siRNA Details
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019309.2 | NP_062182.1 | 3 | 699 | ||
GenBank mRNA | Z11548.1 | - | 719 | |||
Z11715.1 | - | 3 | 497 |

Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019309.2 | NP_062182.1 | 3 | 699 | ||
GenBank mRNA | Z11548.1 | - | 719 | |||
Z11715.1 | - | 3 | 497 |