This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CLTC SIRNA Assays ›
Gene Symbol:
Cltc
Gene Name:
clathrin, heavy chain (Hc)
Gene Aliases:
-
Chromosome Location:
Chr.10: 74014560 - 74070575 on Build Rnor_6.0
UniGene ID:
Rn.3589
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 50806

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cltc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019299.1 NP_062172.1 2 253
XM_006247107.2 XP_006247169.1 2 562
XM_006247108.2 XP_006247170.1 2 561
XM_006247109.2 XP_006247171.1 2 561
GenBank mRNA J03583.1 - 2 253

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
intracellular protein transport
receptor-mediated endocytosis
Golgi organization
mitotic nuclear division
vesicle-mediated transport
receptor internalization
transferrin transport
negative regulation of hyaluronan biosynthetic process
negative regulation of protein localization to plasma membrane
double-stranded RNA binding
structural molecule activity
protein binding
protein C-terminus binding
protein kinase binding
ankyrin binding
heat shock protein binding
clathrin light chain binding
peptide binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0