This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FANCE SIRNA Assays ›
Gene Symbol:
Fance
Gene Name:
Fanconi anemia, complementation group E
Gene Aliases:
2810451D06Rik, AI415634, AW209126
Chromosome Location:
Chr.17: 28313530 - 28326574 on Build GRCm38
UniGene ID:
Mm.201859
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 507935

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fance
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163819.1 NP_001157291.1 2 678
NM_001163820.1 NP_001157292.1 2 678
GenBank mRNA AK013325.1 - 2 641
AK135017.1 - 3 582
AK172283.1 - 2 662
AK213002.1 - 1 84
BC016538.1 - 2 636
BC039987.1 - 2 641
BY740269.1 - 2 662

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0