This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNMB1 SIRNA Assays ›
Gene Symbol:
Kcnmb1
Gene Name:
potassium channel subfamily M regulatory beta subunit 1
Gene Aliases:
BKbeta, BKbeta1, k(VCA)beta-1, slo-beta, slo-beta-1
Chromosome Location:
Chr.10: 18910811 - 18920125 on Build Rnor_6.0
UniGene ID:
Rn.10820
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 50784

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnmb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019273.1 NP_062146.1 644
XM_006246089.2 XP_006246151.1 2150
XM_008767562.1 XP_008765784.1 3102
XM_008767563.1 XP_008765785.1 2969
XM_008767564.1 XP_008765786.1 3102
GenBank mRNA AB010963.1 - 353
AF020712.1 - 644
FJ154955.1 - 333
U40602.1 - 331
U54498.1 - 382

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_031169, AF020711, AK038987, AJ001291, BC013338.

Gene Ontology Categories:

Function(s) Process(es)

detection of calcium ion
chemical synaptic transmission
response to calcium ion
potassium ion transmembrane transport
calcium-activated potassium channel activity
potassium channel regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0