siRNA Details
Gene Symbol: Cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027468.1 | NP_081744.1 | 9 | 3451 | ||
GenBank mRNA | AK004327.1 | - | 1 | 204 | ||
AK013053.1 | - | 1 | 110 |

Gene Symbol: Cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027468.1 | NP_081744.1 | 9 | 3451 | ||
GenBank mRNA | AK004327.1 | - | 1 | 204 | ||
AK013053.1 | - | 1 | 110 |