This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CPM SIRNA Assays ›
Gene Symbol:
Cpm
Gene Name:
carboxypeptidase M
Gene Aliases:
1110060I01Rik, 5730456K23Rik, AA589379, E030045M14Rik
Chromosome Location:
Chr.10: 117629500 - 117687352 on Build GRCm38
UniGene ID:
Mm.339332
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 507240

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cpm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027468.1 NP_081744.1 9 1610
GenBank mRNA AK017670.1 - 3 840
BC047389.1 - 8 1596
BC055073.1 - 9 1615
BC088735.1 - 9 1615
BC100404.1 - 9 1604 30471377

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
biological_process
molecular_function
carboxypeptidase activity
metallocarboxypeptidase activity
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0