This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHP2 SIRNA Assays ›
Gene Symbol:
Chp2
Gene Name:
calcineurin-like EF hand protein 2
Gene Aliases:
2010110P09Rik, Cbhp2
Chromosome Location:
Chr.7: 122208621 - 122222539 on Build GRCm38
UniGene ID:
Mm.96997
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 507202

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027363.1 NP_081639.1 5, 6 622
GenBank mRNA AK008392.1 - 5, 6 456
BC119202.1 - 5, 6 622 8733951
BC119204.1 - 5, 6 622 8733953

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_182738, XM_008759746, XM_008759745, AB086189.

Gene Ontology Categories:

Function(s) Process(es)

transport
sodium ion transport
regulation of pH
positive regulation of cell proliferation
positive regulation of phosphatase activity
protein transport
positive regulation of protein import into nucleus
positive regulation of transcription from RNA polymerase II promoter
positive regulation of calcineurin-NFAT signaling cascade
cellular response to calcium ion
calcium ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0