This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EML1 SIRNA Assays ›
Gene Symbol:
Eml1
Gene Name:
echinoderm microtubule associated protein like 1
Gene Aliases:
1110008N23Rik, A930030P13Rik, AA171013, AI847476, AI853955, ELP79, EMAP, EMAPL
Chromosome Location:
Chr.12: 108371153 - 108539564 on Build GRCm38
UniGene ID:
Mm.236645
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 506668

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eml1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001043335.1 NP_001036800.1 9 1307
NM_001043336.2 NP_001036801.1 9 924
NM_001286346.1 NP_001273275.1 9 1061
NM_001286347.1 NP_001273276.1 10 975
GenBank mRNA AK044662.1 - 9 1046
BC053094.1 - 9 1141 6400458
BC059839.1 - 9 902 6417403
BC079582.1 - 9 960 5702743
BC098329.1 - 8 703
BC125289.1 - 10 946 40129804
BC144757.1 - 10 946
BC150975.1 - 9 1061 9087888

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
hematopoietic progenitor cell differentiation
mitotic spindle organization
neuroblast proliferation
brain development
microtubule binding
tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0