This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYP4A31 SIRNA Assays ›
Gene Symbol:
Cyp4a31
Gene Name:
cytochrome P450, family 4, subfamily a, polypeptide 31
Gene Aliases:
-
Chromosome Location:
Chr.4: 115563626 - 115579015 on Build GRCm38
UniGene ID:
Mm.482086
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 506249

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyp4a31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252539.1 NP_001239468.1 11 1560
NM_201640.3 NP_964002.2 12 1622
GenBank mRNA BC013476.1 - 11 1511 4237837

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

leukotriene B4 catabolic process
oxidation-reduction process
arachidonic acid monooxygenase activity
arachidonic acid epoxygenase activity
alkane 1-monooxygenase activity
leukotriene-B4 20-monooxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0