This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD47 SIRNA Assays ›
Gene Symbol:
Cd47
Gene Name:
Cd47 molecule
Gene Aliases:
Itgp
Chromosome Location:
Chr.11: 53511485 - 53575754 on Build Rnor_6.0
UniGene ID:
Rn.7409
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 50543

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd47
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019195.2 NP_062068.1 2 484
XM_006248262.2 XP_006248324.1 2 1063
XM_006248263.2 XP_006248325.1 2 1063
XM_006248264.2 XP_006248326.1 2 1063
XM_006248265.2 XP_006248327.1 2 1063
XM_006248266.2 XP_006248328.1 2 506
XM_006248267.2 XP_006248329.1 2 1063
XM_006248268.2 XP_006248330.1 2 1261
GenBank mRNA AF017437.1 - 2 370
BC085740.1 - 2 484 7189894
D87659.1 - 2 394
FQ209533.1 - 1 107
FQ232318.1 - 1 107

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010581, AK151922, AB012693, Z25524, BC009094, and others.

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
integrin-mediated signaling pathway
opsonization
positive regulation of cell proliferation
response to bacterium
cell migration
positive regulation of cell-cell adhesion
positive regulation of inflammatory response
positive regulation of phagocytosis
positive regulation of T cell activation
thrombospondin receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0