This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SEPP1 SIRNA Assays ›
Gene Symbol:
Sepp1
Gene Name:
selenoprotein P, plasma, 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 53105912 - 53116198 on Build Rnor_6.0
UniGene ID:
Rn.1451
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 50540

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sepp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083911.1 NP_001077380.1 2, 3 349
NM_019192.2 NP_062065.2 2, 3 277
GenBank mRNA BC059137.1 - 2, 3 278
BC072539.1 - 2, 3 266 7104195
CB699091.1 - 2, 3 349
D25221.1 - 2, 3 258
DN933732.1 - 2, 3 597
M63574.1 - 237

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009155, NM_001042613, NM_001042614, AK159269, AK159524, and others.

Gene Ontology Categories:

Function(s) Process(es)

selenium compound metabolic process
brain development
locomotory behavior
post-embryonic development
response to selenium ion
sexual reproduction
growth
selenium binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0