This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLF14 SIRNA Assays ›
Gene Symbol:
Klf14
Gene Name:
Kruppel-like factor 14
Gene Aliases:
5330411L03Rik, AL022736, BTEB5
Chromosome Location:
Chr.6: 30956021 - 30958990 on Build GRCm38
UniGene ID:
Mm.204671
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 505380

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klf14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135093.1 NP_001128565.1 1 1941
GenBank mRNA AK030435.1 - 2 1235

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sphingolipid mediated signaling pathway
transcriptional activator activity, RNA polymerase II transcription factor binding
nucleic acid binding
DNA binding
chromatin binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0