This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM5460 SIRNA Assays ›
Gene Symbol:
Gm5460
Gene Name:
predicted gene 5460
Gene Aliases:
EG432838
Chromosome Location:
Chr.14: 34016292 - 34047114 on Build GRCm38
UniGene ID:
Mm.318383
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 504558

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm5460
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034880.2 NP_001030052.1 5 1831
GenBank mRNA AK142650.1 - 5 1832
BC150959.1 - 5 1795 9087870
BC151174.1 - 5 1795 9088063

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0