This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2U SIRNA Assays ›
Gene Symbol:
Ube2u
Gene Name:
ubiquitin-conjugating enzyme E2U (putative)
Gene Aliases:
-
Chromosome Location:
Chr.4: 100478867 - 100550145 on Build GRCm38
UniGene ID:
Mm.318133
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 504148

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ube2u
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033773.4 NP_001028945.2 1 336
GenBank mRNA AK133379.1 - 1 337
BB616918.1 - 1 337
BC150880.1 - 1 328 9087791
BC150888.1 - 1 328 9087799
BC172031.1 - 1 328 9054317

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
DNA repair
biological_process
histone ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
molecular_function
ubiquitin protein ligase binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0