This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBLN5 SIRNA Assays ›
Gene Symbol:
Fbln5
Gene Name:
fibulin 5
Gene Aliases:
-
Chromosome Location:
Chr.6: 125644797 - 125723957 on Build Rnor_6.0
UniGene ID:
Rn.1699
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 50411

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbln5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019153.3 NP_062026.2 3, 4 345
XM_006240459.2 XP_006240521.1 3, 4 576
GenBank mRNA AF112153.1 - 3, 4 469
AF137350.1 - 3, 4 272
BC078845.1 - 3, 4 345 7123752

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006329, AY358898, AK307592, AK075147, AF112152, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
cell adhesion
extracellular matrix organization
protein localization to cell surface
secretion
elastic fiber assembly
regulation of removal of superoxide radicals
integrin binding
calcium ion binding
protein binding
protein C-terminus binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0