This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP703 SIRNA Assays ›
Gene Symbol:
Zfp703
Gene Name:
zinc finger protein 703
Gene Aliases:
1110032O19Rik, AI430822, AL022941, Csmn1, End2, Zeppo1, Znf703, Zpo1
Chromosome Location:
Chr.8: 26977336 - 26981462 on Build GRCm38
UniGene ID:
Mm.212469
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 503925

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp703
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101502.1 NP_001094972.1 2 2730
NM_001110508.1 NP_001103978.1 3 2864
GenBank mRNA AK027951.1 - 1 105
BB609653.1 - 1 105
CD352751.1 - 1 512

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109425.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell proliferation
positive regulation of epithelial to mesenchymal transition
regulation of transforming growth factor beta receptor signaling pathway
positive regulation of cell migration
positive regulation of mammary gland epithelial cell proliferation
negative regulation of homotypic cell-cell adhesion
adherens junction assembly
negative regulation of transcription, DNA-templated
regulation of cell cycle
mammary gland epithelial cell differentiation
regulation of canonical Wnt signaling pathway
cellular response to estradiol stimulus
nucleic acid binding
metal ion binding
repressing transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0