This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLFR221 SIRNA Assays ›
Gene Symbol:
Olfr221
Gene Name:
olfactory receptor 221
Gene Aliases:
MOR205-1, MOR205-2, Olfr1514, Olfr221-ps1
Chromosome Location:
Chr.14: 52035174 - 52036109 on Build GRCm38
UniGene ID:
Mm.377946
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 503157

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr221
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001808.2 NP_001001808.2 1 595
GenBank mRNA BC147427.1 - 1 629 9056495
BC147428.1 - 1 629 9056478
BC151091.1 - 1 901 9088004
BC151095.1 - 1 894 9088008

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000834, XM_008770559.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0