This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AVPR1B SIRNA Assays ›
Gene Symbol:
Avpr1b LOC100909648
Gene Name:
arginine vasopressin receptor 1B
vasopressin V1b receptor-like
Gene Aliases:
-
-
Chromosome Location:
Chr.13: 48367307 - 48379134 on Build Rnor_6.0
Chromosome Location:
Chr.13: 48390434 - 48400768 on Build Rnor_6.0
UniGene ID:
Rn.10096
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 50064

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Avpr1b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017205.3 NP_058901.3 3 1860
XM_008769445.1 XP_008767667.1 3 1538
GenBank mRNA AF064541.1 - 2 560
D45400.1 - 2 1389
U27322.1 - 2 1753
Gene Symbol: LOC100909648
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289800.1 NP_001276729.1 2 1328
XM_008769404.1 XP_008767626.1 3 1531

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of systemic arterial blood pressure by vasopressin
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
positive regulation of glutamate secretion
positive regulation of cellular pH reduction
cellular response to hormone stimulus
hyperosmotic salinity response
positive regulation of vasoconstriction
response to peptide
G-protein coupled receptor activity
vasopressin receptor activity
peptide hormone binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0