siRNA Details
Gene Symbol: Gm19668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205034.1 | NP_001191963.1 | 1 | 184 |

Gene Symbol: Gm19668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205034.1 | NP_001191963.1 | 1 | 184 |