This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other INSR SIRNA Assays ›
Gene Symbol:
Insr
Gene Name:
insulin receptor
Gene Aliases:
-
Chromosome Location:
Chr.12: 1678620 - 1816587 on Build Rnor_6.0
UniGene ID:
Rn.9876
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 49576

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Insr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017071.2 NP_058767.2 2 493
XM_006248752.2 XP_006248814.1 2 666
XM_006248753.2 XP_006248815.1 2 666
GenBank mRNA KF112847.1 - 2 219
KF112848.1 - 2 219
M29014.1 - 2 489

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
heart morphogenesis
regulation of transcription, DNA-templated
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of cell proliferation
insulin receptor signaling pathway
epidermis development
response to hormone
response to glucose
organ morphogenesis
response to manganese ion
regulation of hydrogen peroxide metabolic process
positive regulation of glycoprotein biosynthetic process
negative regulation of gene expression
response to activity
peptidyl-tyrosine phosphorylation
transformation of host cell by virus
signal transduction by protein phosphorylation
male sex determination
positive regulation of cell migration
exocrine pancreas development
response to nutrient levels
activation of protein kinase activity
activation of protein kinase B activity
response to estradiol
negative regulation of transporter activity
response to insulin
cellular response to insulin stimulus
response to vitamin D
response to testosterone
response to tumor necrosis factor
peptidyl-tyrosine autophosphorylation
positive regulation of phosphorylation
glucose homeostasis
positive regulation of MAPK cascade
positive regulation of nitric oxide biosynthetic process
fat cell differentiation
response to ethanol
positive regulation of glycogen biosynthetic process
positive regulation of DNA replication
positive regulation of glycolytic process
positive regulation of mitotic nuclear division
regulation of embryonic development
positive regulation of glucose import
protein autophosphorylation
positive regulation of developmental growth
protein heterotetramerization
response to glucocorticoid
positive regulation of protein kinase B signaling
positive regulation of respiratory burst
cellular response to growth factor stimulus
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
insulin-activated receptor activity
insulin-like growth factor receptor binding
protein binding
ATP binding
GTP binding
protein kinase binding
protein phosphatase binding
protein domain specific binding
lipoic acid binding
insulin-like growth factor I binding
insulin-like growth factor II binding
protein complex binding
3-phosphoinositide-dependent protein kinase binding
phosphatidylinositol 3-kinase binding
insulin binding
insulin receptor substrate binding
PTB domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0