This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LDHA SIRNA Assays ›
Gene Symbol:
Ldha
Gene Name:
lactate dehydrogenase A
Gene Aliases:
Ldh1
Chromosome Location:
Chr.1: 102900288 - 102909713 on Build Rnor_6.0
UniGene ID:
Rn.107896
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 49532

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ldha
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017025.1 NP_058721.1 7, 8 945
XM_006229232.2 XP_006229294.1 7, 8 1113
GenBank mRNA BC060587.1 - 7, 8 897 6919582
BC084698.1 - 6, 7 847 7122339
BC168737.1 - 7, 8 944 6917937
FQ214854.1 - 7, 8 948
M54926.1 - 23
X01964.1 - 7, 8 945

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
lactate metabolic process
response to nutrient
response to glucose
response to organic cyclic compound
NAD metabolic process
substantia nigra development
cellular response to extracellular stimulus
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
response to estrogen
cellular carbohydrate metabolic process
post-embryonic organ development
response to cAMP
oxidation-reduction process
lactate dehydrogenase activity
L-lactate dehydrogenase activity
protein binding
kinase binding
identical protein binding
NAD binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0