This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACADM SIRNA Assays ›
Gene Symbol:
Acadm
Gene Name:
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases:
MCAD
Chromosome Location:
Chr.2: 260124418 - 260148589 on Build Rnor_6.0
UniGene ID:
Rn.6302
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 49500

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Acadm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016986.2 NP_058682.2 10 1035
GenBank mRNA FQ214755.1 - 4 363
J02791.1 - 10 950

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

liver development
glycogen biosynthetic process
organic acid metabolic process
regulation of gluconeogenesis
fatty acid beta-oxidation
heart development
response to nutrient
response to cold
carnitine metabolic process
response to hormone
post-embryonic development
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to drug
response to starvation
carnitine biosynthetic process
response to copper ion
protein homotetramerization
response to glucocorticoid
medium-chain fatty acid metabolic process
medium-chain fatty acid catabolic process
cardiac muscle cell differentiation
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
isomerase activity
identical protein binding
flavin adenine dinucleotide binding
medium-chain-acyl-CoA dehydrogenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0