This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF136 SIRNA Assays ›
Gene Symbol:
ZNF136
Gene Name:
zinc finger protein 136
Gene Aliases:
pHZ-20
Chromosome Location:
Chr.19: 12162672 - 12189880 on Build GRCh38
UniGene ID:
Hs.479874
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 4935

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF136
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003437.3 NP_003428.1 4 2296
XM_011528269.2 XP_011526571.1 5 2791
XM_017027243.1 XP_016882732.1 3 2554
GenBank mRNA AK123008.1 - 3 3313
AY500356.1 - 1 810
BC006421.1 - 4 2249 4040430
U09367.1 - 4 2223

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
DNA binding
transcription corepressor activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0