This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ROS1 SIRNA Assays ›
Gene Symbol:
Ros1
Gene Name:
ROS proto-oncogene 1 , receptor tyrosine kinase
Gene Aliases:
ROS1C
Chromosome Location:
Chr.20: 33100190 - 33323548 on Build Rnor_6.0
UniGene ID:
Rn.87436
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 48612

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ros1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012874.1 NP_037006.1 3 587
XM_008772862.1 XP_008771084.1 3 768
XM_008772863.1 XP_008771085.1 3 303
GenBank mRNA M35104.1 - 3 587
M35105.1 - 3 587
M35106.1 - 3 587

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

columnar/cuboidal epithelial cell development
protein phosphorylation
spermatogenesis
cell proliferation
negative regulation of gene expression
regulation of phosphate transport
cell growth
peptidyl-tyrosine phosphorylation
signal transduction by protein phosphorylation
cell differentiation
regulation of TOR signaling
peptidyl-tyrosine autophosphorylation
regulation of ERK1 and ERK2 cascade
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ATP binding
protein phosphatase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0