This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD2 SIRNA Assays ›
Gene Symbol:
Cd2
Gene Name:
Cd2 molecule
Gene Aliases:
CD2R, LFA-2, LFA2, OX-34, OX34
Chromosome Location:
Chr.2: 203666706 - 203680073 on Build Rnor_6.0
UniGene ID:
Rn.10328
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 48576

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012830.1 NP_036962.1 2 437
GenBank mRNA BC088164.1 - 2 367 7374969
X05111.1 - 2 437

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

single organismal cell-cell adhesion
positive regulation of tumor necrosis factor production
heterotypic cell-cell adhesion
T cell activation
positive regulation of interferon-gamma secretion
positive regulation of interleukin-8 secretion
glycoprotein binding
antigen binding
receptor activity
receptor binding
protein kinase binding
receptor tyrosine kinase binding
protein homodimerization activity
protein self-association

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0