siRNA Details
Gene Symbol: Gpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012774.1 | NP_036906.1 | 7 | 1559 | ||
GenBank mRNA | BC085756.1 | - | 7 | 1574 | 7122343 | |
M22400.1 | - | 7 | 1559 |

Gene Symbol: Gpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012774.1 | NP_036906.1 | 7 | 1559 | ||
GenBank mRNA | BC085756.1 | - | 7 | 1574 | 7122343 | |
M22400.1 | - | 7 | 1559 |