This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CALCRL SIRNA Assays ›
Gene Symbol:
Calcrl
Gene Name:
calcitonin receptor-like
Gene Aliases:
CLR, Crlr, RATCRLR, RNCLR
Chromosome Location:
Chr.3: 71747940 - 71845487 on Build Rnor_6.0
UniGene ID:
Rn.11202
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 48307

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Calcrl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012717.1 NP_036849.1 6 575
XM_006234438.2 XP_006234500.1 6 830
XM_006234439.2 XP_006234501.1 5 668
GenBank mRNA L27487.1 - 6 848
X70658.1 - 6 575

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cAMP biosynthetic process
calcium ion transport
G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
heart development
positive regulation of cell proliferation
protein transport
positive regulation of cAMP biosynthetic process
receptor internalization
negative regulation of smooth muscle contraction
positive regulation of smooth muscle cell proliferation
negative regulation of inflammatory response
cellular response to sucrose stimulus
adrenomedullin receptor activity
calcitonin gene-related peptide receptor activity
calcitonin receptor activity
protein transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0