This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CKM SIRNA Assays ›
Gene Symbol:
Ckm
Gene Name:
creatine kinase, muscle
Gene Aliases:
Ckmm
Chromosome Location:
Chr.1: 80321585 - 80331841 on Build Rnor_6.0
UniGene ID:
Rn.10756
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 47885

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ckm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012530.2 NP_036662.1 6 834
GenBank mRNA FM053193.1 - 2 150
FQ214594.1 - 6 810
FQ214655.1 - 6 811
FQ214825.1 - 5 631
FQ215410.1 - 6 810
FQ215629.1 - 4 427
FQ216321.1 - 5 662
FQ216507.1 - 6 805
FQ216611.1 - 4 427
FQ217421.1 - 5 675
FQ224848.1 - 5 661
FQ224918.1 - 6 811
M10140.1 - 6 807
M14864.1 - 4 480

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
phosphocreatine biosynthetic process
creatine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header