This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other A2M SIRNA Assays ›
Gene Symbol:
A2m LOC100911545
Gene Name:
alpha-2-macroglobulin
alpha-2-macroglobulin-like
Gene Aliases:
A2MAC1, A2m1, A2maa, Mam
-
Chromosome Location:
Chr.4: 154309426 - 154359138 on Build Rnor_6.0
Chromosome Location:
Chr.4: 154422862 - 154473042 on Build Rnor_6.0
UniGene ID:
Rn.225884
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 47849

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: A2m
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012488.2 NP_036620.2 12 1489
GenBank mRNA AY921651.1 - 12 1448
BC098922.1 - 12 1489 7458629
FQ223246.1 - 12 1511
FQ228339.1 - 12 1688
J02635.1 - 12 1511
M11793.1 - 2 191
Gene Symbol: LOC100911545
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749828.2 XP_003749876.1 1795

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of complement activation, lectin pathway
acute-phase response
inflammatory response
response to nutrient
response to wounding
response to carbon dioxide
negative regulation of endopeptidase activity
stem cell differentiation
response to glucocorticoid
protease binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
receptor binding
growth factor binding
enzyme binding
interleukin-8 binding
interleukin-1 binding
peptidase inhibitor activity
tumor necrosis factor binding
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0