This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TPST1 SIRNA Assays ›
Gene Symbol:
TPST1
Gene Name:
tyrosylprotein sulfotransferase 1
Gene Aliases:
TANGO13A
Chromosome Location:
Chr.7: 66205272 - 66360451 on Build GRCh38
UniGene ID:
Hs.421194
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 4778

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TPST1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003596.3 NP_003587.1 2 503
XM_005250642.2 XP_005250699.1 2 499
XM_011516634.2 XP_011514936.1 2 499
XM_011516635.2 XP_011514937.1 2 499
XM_017012724.1 XP_016868213.1 3 586
XM_017012725.1 XP_016868214.1 3 586
XM_017012726.1 XP_016868215.1 2 499
XM_017012727.1 XP_016868216.1 2 499
GenBank mRNA AF038009.1 - 1 232
AK313098.1 - 2 441
BC013188.1 - 2 478 3918922
CR542060.1 - 1 151
CR542080.1 - 1 151

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

peptidyl-tyrosine sulfation
inflammatory response
protein-tyrosine sulfotransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0