This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MTUS1 SIRNA Assays ›
Gene Symbol:
Mtus1
Gene Name:
microtubule associated tumor suppressor 1
Gene Aliases:
ATIP4, Atip3b, Mtsg1
Chromosome Location:
Chr.16: 54240897 - 54386131 on Build Rnor_6.0
UniGene ID:
Rn.95344
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 47483

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mtus1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178093.2 NP_835194.2 1 335
GenBank mRNA AB190507.1 - 1 412
AY208915.1 - 1 103
BC072537.1 - 1 335
BC166591.1 - 1 384 7938925

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001005865, BE653450, AY626781, AK030510, AY246699, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of macrophage chemotaxis
cellular response to peptide hormone stimulus
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0