This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STK16 SIRNA Assays ›
Gene Symbol:
Stk16
Gene Name:
serine/threonine kinase 16
Gene Aliases:
F52, PKL12
Chromosome Location:
Chr.9: 82411010 - 82414249 on Build Rnor_6.0
UniGene ID:
Rn.13308
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 47384

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stk16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173142.1 NP_775165.1 8 1457
XM_006245160.1 XP_006245222.1 7 1364
GenBank mRNA D86220.1 - 8 1457
FQ213827.1 - 8 1455

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of signal transduction
peptidyl-tyrosine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
cellular response to transforming growth factor beta stimulus
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein kinase activity
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0