This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HMGCS2 SIRNA Assays ›
Gene Symbol:
Hmgcs2
Gene Name:
3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial)
Gene Aliases:
Hmgcs1, Mt3h3mg
Chromosome Location:
Chr.2: 200452619 - 200480785 on Build Rnor_6.0
UniGene ID:
Rn.29594
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 47349

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hmgcs2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173094.2 NP_775117.2 5 1020
XM_006233002.2 XP_006233064.1 5 1049
GenBank mRNA BC078695.1 - 5 1020 7112644
BC083543.1 - 5 1020 7132009
M33648.1 - 5 1038

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

kidney development
liver development
cholesterol biosynthetic process
brain development
midgut development
response to nutrient
isoprenoid biosynthetic process
response to temperature stimulus
response to bacterium
response to metal ion
response to organic cyclic compound
lung development
response to insulin
cellular response to insulin stimulus
cellular response to hormone stimulus
multicellular organismal response to stress
response to testosterone
response to glucagon
response to triglyceride
response to monosaccharide
response to prostaglandin F
response to drug
response to starvation
response to peptide hormone
response to ethanol
ketone body biosynthetic process
response to glucocorticoid
response to cAMP
response to growth hormone
adipose tissue development
response to fatty acid
response to linoleic acid
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to glucocorticoid stimulus
cellular response to fatty acid
cellular response to organic cyclic compound
hydroxymethylglutaryl-CoA synthase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0