This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NPC2 SIRNA Assays ›
Gene Symbol:
Npc2
Gene Name:
Niemann-Pick disease, type C2
Gene Aliases:
re1
Chromosome Location:
Chr.6: 108467410 - 108488330 on Build Rnor_6.0
UniGene ID:
Rn.3491
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 47273

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Npc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173118.2 NP_775141.2 2 266
GenBank mRNA AJ515237.1 - 2 201
BC058132.1 - 2 229 6920130
FQ214040.1 - 2 187
FQ216545.1 - 2 191
FQ219301.1 - 2 208
FQ219863.1 - 2 199
FQ220660.1 - 2 187
FQ220919.1 - 2 193
FQ228456.1 - 2 191
FQ229422.1 - 2 246
FQ229782.1 - 2 187
FQ229783.1 - 2 189
FQ230107.1 - 2 193

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023409, AK167869, AK012239, BC007190, AK153203, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
response to virus
cholesterol transport
intracellular sterol transport
intracellular cholesterol transport
cholesterol efflux
cholesterol homeostasis
molecular_function
cholesterol binding
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0