This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR3 SIRNA Assays ›
Gene Symbol:
Gpr3
Gene Name:
G protein-coupled receptor 3
Gene Aliases:
Gpcr3
Chromosome Location:
Chr.5: 151394512 - 151397624 on Build Rnor_6.0
UniGene ID:
Rn.83629
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 47238

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpr3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153727.1 NP_714949.1 1 420
XM_006239006.2 XP_006239068.1 2 559
GenBank mRNA AJ427482.1 - 1 420

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
positive regulation of cAMP metabolic process
regulation of meiotic nuclear division
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0