This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAPRE1 SIRNA Assays ›
Gene Symbol:
Mapre1
Gene Name:
microtubule-associated protein, RP/EB family, member 1
Gene Aliases:
Eb1
Chromosome Location:
Chr.3: 149221353 - 149249469 on Build Rnor_6.0
UniGene ID:
Rn.7652
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 47112

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mapre1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138509.3 NP_612518.2 2 190
XM_006235294.2 XP_006235356.1 2 173
XM_006235295.2 XP_006235357.1 2 243
GenBank mRNA BC081726.1 - 2 190 7192469
U75920.1 - 2 158

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic nuclear division
regulation of microtubule polymerization
negative regulation of microtubule polymerization
protein localization to microtubule
cell division
positive regulation of microtubule plus-end binding
poly(A) RNA binding
microtubule plus-end binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0