This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FZD1 SIRNA Assays ›
Gene Symbol:
Fzd1
Gene Name:
frizzled class receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.4: 26470864 - 26475262 on Build Rnor_6.0
UniGene ID:
Rn.6575
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 46822

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fzd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021266.3 NP_067089.2 1 1771

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
membranous septum morphogenesis
muscular septum morphogenesis
outflow tract morphogenesis
G-protein coupled receptor signaling pathway
cell-cell signaling
Wnt signaling pathway
negative regulation of BMP signaling pathway
epithelial cell differentiation
negative regulation of catenin import into nucleus
autocrine signaling
non-canonical Wnt signaling pathway
response to drug
canonical Wnt signaling pathway involved in mesenchymal stem cell differentiation
canonical Wnt signaling pathway involved in osteoblast differentiation
regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
lung alveolus development
positive regulation of sequence-specific DNA binding transcription factor activity
hard palate development
canonical Wnt signaling pathway
ventricular septum morphogenesis
cellular response to vitamin D
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
negative regulation of canonical Wnt signaling pathway
planar cell polarity pathway involved in neural tube closure
G-protein coupled receptor activity
receptor binding
frizzled binding
protein binding
Wnt-protein binding
PDZ domain binding
identical protein binding
protein homodimerization activity
Wnt-activated receptor activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0