This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STX2 SIRNA Assays ›
Gene Symbol:
Stx2
Gene Name:
syntaxin 2
Gene Aliases:
Epim
Chromosome Location:
Chr.12: 31324047 - 31360249 on Build Rnor_6.0
UniGene ID:
Rn.10623
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 46776

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stx2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012748.1 NP_036880.1 4 252
XM_006249249.2 XP_006249311.1 4 410
XM_006249250.2 XP_006249312.1 4 410
XM_006249253.2 XP_006249315.1 5 2156
XM_008769188.1 XP_008767410.1 5 2623
GenBank mRNA AY302700.1 - 4 252
L20823.1 - 4 252
L20888.1 - 4 252
L20889.1 - 4 252

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007941, NM_001286034, NM_001286033, BC046279, AK165552, and others.

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle targeting
vesicle fusion
acrosome reaction
regulation of gene expression
vesicle-mediated transport
microvillus assembly
cell differentiation
regulation of blood coagulation
epithelial cell differentiation
synaptic vesicle fusion to presynaptic active zone membrane
response to hydroperoxide
intracellular transport
vesicle docking
digestive tract morphogenesis
protein oligomerization
regulation of cytoskeleton organization
membrane fusion
SNARE binding
SNAP receptor activity
protein binding
protein dimerization activity
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0