This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ISG15 SIRNA Assays ›
Gene Symbol:
ISG15
Gene Name:
ISG15 ubiquitin-like modifier
Gene Aliases:
G1P2, IFI15, IMD38, IP17, UCRP, hUCRP
Chromosome Location:
Chr.1: 1013467 - 1014540 on Build GRCh38
UniGene ID:
Hs.458485
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 46107

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ISG15
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005101.3 NP_005092.1 2 552
GenBank mRNA AY168648.1 - 1 445
BC009507.2 - 2 486 3545944
BT007297.1 - 1 445
BU161226.1 - 2 509
M13755.1 - 2 520

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

modification-dependent protein catabolic process
translesion synthesis
negative regulation of protein ubiquitination
ISG15-protein conjugation
negative regulation of type I interferon production
regulation of interferon-gamma production
response to type I interferon
defense response to bacterium
negative regulation of viral genome replication
positive regulation of erythrocyte differentiation
defense response to virus
type I interferon signaling pathway
protein binding
protein tag

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0