This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYP2C9 SIRNA Assays ›
Gene Symbol:
CYP2C9
Gene Name:
cytochrome P450 family 2 subfamily C member 9
Gene Aliases:
CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9
Chromosome Location:
Chr.10: 94905867 - 94989395 on Build GRCh38
UniGene ID:
Hs.282624
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 45948

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CYP2C9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000771.3 NP_000762.2 1 165
XM_017015758.1 XP_016871247.1 2 585
GenBank mRNA AK289420.1 - 1 165
AK298458.1 - 1 165
BC020754.1 - 1 166
BC070317.1 - 1 162
BC125054.1 - 1 140 40116637
BJ993098.1 - 1 165
D00173.1 - 1 131
M15331.1 - 1 125
M21939.1 - 1 126
M61855.1 - 1 152
M61857.1 - 1 150
S46963.1 - 1 103

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

xenobiotic metabolic process
steroid metabolic process
monoterpenoid metabolic process
drug metabolic process
epoxygenase P450 pathway
urea metabolic process
monocarboxylic acid metabolic process
drug catabolic process
exogenous drug catabolic process
cellular amide metabolic process
oxidation-reduction process
oxidative demethylation
omega-hydroxylase P450 pathway
cholesterol 25-hydroxylase activity
monooxygenase activity
iron ion binding
drug binding
arachidonic acid epoxygenase activity
steroid hydroxylase activity
oxidoreductase activity
(S)-limonene 6-monooxygenase activity
(S)-limonene 7-monooxygenase activity
oxygen binding
heme binding
caffeine oxidase activity
(R)-limonene 6-monooxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0