This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other METTL5 SIRNA Assays ›
Gene Symbol:
METTL5
Gene Name:
methyltransferase like 5
Gene Aliases:
HSPC133
Chromosome Location:
Chr.2: 169811757 - 169824931 on Build GRCh38
UniGene ID:
Hs.470553
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 45207

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: METTL5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293186.1 NP_001280115.1 2 270
NM_001293187.1 NP_001280116.1 2 184
NM_014168.3 NP_054887.2 1 411
XM_011511023.1 XP_011509325.1 2 211
GenBank mRNA AF161482.1 - 2 159
AF201938.1 - 2 228
AL523293.3 - 1 393
BC000921.2 - 1 323 3445582
BC093014.1 - 2 166 4799279
CA337169.1 - 2 185

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

methylation
nucleic acid binding
methyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0